1,1,1,3,3,3-Hexafluoroisopropyl methacrylate 98%
Leverantör: Apollo Scientific
Fara
Synonyms: Hexafluoroisopropyl methacrylate, 1,1,1,3,3,3-Hexafluoroisopropylmethacrylate
PC4756-100G PC4756-50G PC4756-250G
APOSPC4756-100GEA 2425 SEK
APOSPC4756-100G APOSPC4756-50G APOSPC4756-250G
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate 98%
1,1,1,3,3,3-Hexafluoroisopropyl methacrylate
Affords low refractive index polymer.
Formel: C₇H₆F₆O₂
MW: 236,12 g/mol
Kokpunkt: 99 °C
Densitet: 1,302 g/cm³ (25 °C)
Flampunkt: 14 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040105
CAS nummer: 3063-94-3
EINECS: 221-309-9
UN: 3272
ADR: 3,II