1,1,1,3,3,3-Hexafluoroisopropyl acrylate 98%

Leverantör: Apollo Scientific
Fara

Synonyms: 1,1,1,3,3,3-Hexafluoropropan-2-yl acrylate

PC4754-100G PC4754-10G PC4754-25G
APOSPC4754-100GEA 3025 SEK
APOSPC4754-100G APOSPC4754-10G APOSPC4754-25G
1,1,1,3,3,3-Hexafluoroisopropyl acrylate 98%
1,1,1,3,3,3-Hexafluoroisopropyl acrylate
Formel: C₆H₄F₆O₂
MW: 222,09 g/mol
Kokpunkt: 31,6…32.4 °C
Densitet: 1,33 g/cm³ (25 °C)
Flampunkt: 10,5 °C
Storage Temperature: Omgivande
MDL Number: MFCD00040104
CAS nummer: 2160-89-6
UN: 3272
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR