1,1,1,4,4,4-Hexafluorobutane 98%

Leverantör: Apollo Scientific

PC4652-100G PC4652-25G
APOSPC4652-100GEA 16675 SEK
APOSPC4652-100G APOSPC4652-25G
1,1,1,4,4,4-Hexafluorobutane 98%
1,1,1,4,4,4-Hexafluorobutane
Formel: C₄H₄F₆
Kokpunkt: 25 °C
Densitet: 1,37 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00236686
CAS nummer: 407-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR