2-(Trifluoromethoxy)phenylboronic acid
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Trifluoromethoxyphenylboronic acid, 2-(Trifluoromethoxy)benzeneboronic acid, (2-(Trifluoromethoxy)phenyl)metaboric acid, Dihydroxy(2-(trifluoromethoxy)phenyl)borane
PC4560-25G PC4560-5G PC4560-100G
APOSPC4560-25GEA 600 SEK
APOSPC4560-25G APOSPC4560-5G APOSPC4560-100G
2-(Trifluoromethoxy)phenylboronic acid
2-(Trifluoromethoxy)phenylboronic acid
Formel: C₇H₆BF₃O₃
MW: 205.93 g/mol
Smältpunkt: 113…120 °C
Flampunkt: >110 °C
Storage Temperature: Kylskåp
MDL Number: MFCD01320763
CAS nummer: 175676-65-0