Fluroxene 97%

Leverantör: Apollo Scientific
Fara

Synonyms: 2,2,2-Trifluoroethyl vinyl ether

PC4526-1G PC4526-100MG PC4526-250MG
APOSPC4526-1GEA 6100 SEK
APOSPC4526-1G APOSPC4526-100MG APOSPC4526-250MG
Fluroxene 97%
Fluroxene
Formel: C₄H₅F₃O
Kokpunkt: 43…45 °C
Densitet: 1,135 g/cm³ (25 °C)
Flampunkt: –29 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00864407
CAS nummer: 406-90-6
EINECS: 206-977-1
UN: 3271
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR