1H,1H,13H-Tetracosafluoro-1-tridecanol 95%

Leverantör: Apollo Scientific
Varning

Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13-Tetracosafluoro-1-tridecanol, 1H,1H,13H-Perfluorotridecan-1-ol, 1H,1H,13H-Perfluoro-1-tridecanol

PC4520-1G PC4520-25G PC4520-5G
APOSPC4520-1GEA 275 SEK
APOSPC4520-1G APOSPC4520-25G APOSPC4520-5G
1H,1H,13H-Tetracosafluoro-1-tridecanol 95%
1H,1H,13H-Tetracosafluoro-1-tridecanol
Formel: C₁₃H₄OF₂₄
MW: 632,14 g/mol
Smältpunkt: 129…130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00155920
CAS nummer: 423-72-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR