Methyl-5-bromo-2-fluorobenzoate
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 5-bromo-2-fluorobenzoate
PC450245-1G PC450245-25G PC450245-5G
APOSPC450245-1GEA 635 SEK
APOSPC450245-1G APOSPC450245-25G APOSPC450245-5G
Methyl-5-bromo-2-fluorobenzoate
Methyl-5-bromo-2-fluorobenzoate
Formel: C₈H₆BrFO₂
MW: 233,0300 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06203710
CAS nummer: 57381-59-6