1,1,1,3,5,5,5-Heptafluoropentan-2,4-dione 97%

Leverantör: Apollo Scientific
Fara

PC4499G-1G PC4499G-5G
APOSPC4499G-1GEA 4050 SEK
APOSPC4499G-1G APOSPC4499G-5G
1,1,1,3,5,5,5-Heptafluoropentan-2,4-dione 97%
1,1,1,3,5,5,5-Heptafluoropentan-2,4-dione
Formel: C₅HF₇O₂
MW: 226,05 g/mol
Kokpunkt: 70…72 °C
Storage Temperature: Omgivande
MDL Number: MFCD00153187
CAS nummer: 77968-17-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR