1-(Heptafluorobutyryl)imidazole 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 1-(Perfluorobutanoyl)-1H-imidazole, N-Heptafluorobutyrylimidazole
PC4494-100G PC4494-5G
APOSPC4494-100GEA 6775 SEK
APOSPC4494-100G APOSPC4494-5G
1-(Heptafluorobutyryl)imidazole 97%
1-(Heptafluorobutyryl)imidazole
Reagent for the derivatisation of amines and alcohols for GC analysis.
Formel: C₇H₃F₇N₂O
MW: 264,10 g/mol
Kokpunkt: 158…163 °C
Smältpunkt: 41548 °C
Densitet: 1,49 g/cm³ (25 °C)
Flampunkt: 77 °C
Storage Temperature: Frys
MDL Number: MFCD00014503
CAS nummer: 32477-35-3
EINECS: 251-063-8