Heptafluorosmörsyra 98%
Leverantör: Apollo Scientific
Fara
Synonyms: Perfluorobutyric acid, HFBA, Heptafluorobutanoic acid
PC4450-100G PC4450-250G PC4450-25G
APOSPC4450-100GEA 1200 SEK
APOSPC4450-100G APOSPC4450-250G APOSPC4450-25G
Heptafluorosmörsyra 98%
Heptafluorosmörsyra
Biochemistry, 20, 4530 (1981): anion-pairing agent for reverse phase HPLC purification of proteins & peptides.
Formel: C₄HF₇O₂
MW: 214,04 g/mol
Kokpunkt: 119…121 °C
Smältpunkt: –18 °C
Densitet: 1,645 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004171
CAS nummer: 375-22-4
EINECS: 206-786-3
UN: 3265
ADR: 8,II