2,2,3,3,4,4,4-Heptafluoro-1-butanol 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 1H,1H-Perfluoro-1-butanol, 2,2,3,3,4,4,4-Heptafluorobutan-1-ol, Heptafluoro-1-butanol
PC4390-100G PC4390-25G
APOSPC4390-100GEA 2700 SEK
APOSPC4390-100G APOSPC4390-25G
2,2,3,3,4,4,4-Heptafluoro-1-butanol 98%
2,2,3,3,4,4,4-Heptafluoro-1-butanol
Intermediate for speciality coating systems; acrylate and methacrylate esters are available.
Formel: C₄H₃F₇O
MW: 200,06 g/mol
Kokpunkt: 95…97 °C
Densitet: 1,6 g/cm³ (25 °C)
Flampunkt: 25 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004674
CAS nummer: 375-01-9
EINECS: 206-782-1
UN: 1987
ADR: 3,III