4'-Fluoro-2'-(trifluoromethyl)propiophenone 97%
Leverantör: Apollo Scientific
Varning
PC4377U-1G PC4377U-250MG
APOSPC4377U-1GEA 2075 SEK
APOSPC4377U-1G APOSPC4377U-250MG
4'-Fluoro-2'-(trifluoromethyl)propiophenone 97%
4'-Fluoro-2'-(trifluoromethyl)propiophenone
Formel: C₁₀H₈F₄O
MW: 220,17 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00061263
CAS nummer: 239107-26-7