(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid
PC4376X-10G PC4376X-1G PC4376X-5G PC4376X-25G
APOSPC4376X-10GEA 2150 SEK
APOSPC4376X-10G APOSPC4376X-1G APOSPC4376X-5G APOSPC4376X-25G
(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid 98%
(4-Fluoro-2-(trifluoromethyl)phenyl)acetic acid
Formel: C₉H₆F₄O₂
MW: 222,14 g/mol
Smältpunkt: 83…86 °C
Storage Temperature: Omgivande
MDL Number: MFCD00061189
CAS nummer: 195447-80-4