(2-Fluoro-3-(trifluoromethyl)phenyl)acetic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Fluoro-3-(trifluoromethyl)phenylacetic acid
PC4376S-1G PC4376S-5G PC4376S-100MG
APOSPC4376S-1GEA 910 SEK
APOSPC4376S-1G APOSPC4376S-5G APOSPC4376S-100MG
(2-Fluoro-3-(trifluoromethyl)phenyl)acetic acid 97%
(2-Fluoro-3-(trifluoromethyl)phenyl)acetic acid
Formel: C₉H₆F₄O₂
MW: 222,14 g/mol
Smältpunkt: 113…116 °C
Storage Temperature: Omgivande
MDL Number: MFCD00061185
CAS nummer: 194943-83-4