2-Fluoro-5-(trifluoromethyl)benzophenone 97%
Leverantör: Apollo Scientific
Varning
Synonyms: (2-Fluoro-5-(trifluoromethyl)phenyl)(phenyl)methanone
PC4374G-1G PC4374G-25G PC4374G-5G
APOSPC4374G-1GEA 260 SEK
APOSPC4374G-1G APOSPC4374G-25G APOSPC4374G-5G
2-Fluoro-5-(trifluoromethyl)benzophenone 97%
2-Fluoro-5-(trifluoromethyl)benzophenone
Formel: C₁₄H₈F₄O
MW: 268.21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00061254
CAS nummer: 199292-40-5