Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: (S)-Isopropyl 2-(((S)-(perfluorophenoxy)(phenoxy)phosphoryl)amino)propanoate
PC43513-100G PC43513-25G PC43513-500G
APOSPC43513-100GEA 1600 SEK
APOSPC43513-100G APOSPC43513-25G APOSPC43513-500G
Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate 98%
Propan-2-yl (2S)-2-{[(S)-pentafluorophenoxy(phenoxy)phosphoryl]amino}propanoate
Formel: C₁₈H₁₇F₅NO₅
MW: 453,298 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD22665799
CAS nummer: 1334513-02-8