6-(4-Fluorophenyl)-3-pyridineboronic acid
Leverantör: Apollo Scientific
Synonyms: 6-(4-Fluorophenyl)pyridine-3-boronic acid
PC412208-100MG PC412208-250MG
APOSPC412208-100MGEA 4200 SEK
APOSPC412208-100MG APOSPC412208-250MG
6-(4-Fluorophenyl)-3-pyridineboronic acid
6-(4-Fluorophenyl)-3-pyridineboronic acid
Formel: C₁₁H₉BFNO₂
MW: 217.01 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09037487
CAS nummer: 1072944-20-7