(±)-2,5-Difluoromandelic acid
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
(±)-2,5-Difluorophenylglycolic acid
PC4114-25G
PC4114-5G
APOSPC4114-25GEA
2950
SEK
APOSPC4114-25G
APOSPC4114-5G
(±)-2,5-Difluoromandelic acid
(±)-2,5-Difluoromandelic acid
Formel:
C₈H₆F₂O₃ MW: 188,13 g/mol Smältpunkt: 110…112 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00061297 CAS nummer: 207853-61-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...