(E)-3-(Dimethylamino)-1-(6-(trifluoromethyl)pyridin-3-yl)prop-2-en-1-one 95+%
Leverantör: Apollo Scientific
Synonyms: 3-(Dimethylamino)-1-(6-(trifluoromethyl)pyridin-3-yl)prop-2-en-1-one
PC409522-1G PC409522-250MG
APOSPC409522-1GEA 5075 SEK
APOSPC409522-1G APOSPC409522-250MG
(E)-3-(Dimethylamino)-1-(6-(trifluoromethyl)pyridin-3-yl)prop-2-en-1-one 95+%
(E)-3-(Dimethylamino)-1-(6-(trifluoromethyl)pyridin-3-yl)prop-2-en-1-one
Formel: C₁₁H₁₁F₃N₂O
MW: 244,22 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD12401613
CAS nummer: 1015078-44-0