3-[4-(Trifluoromethyl)phenyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine 95+%
Leverantör: Apollo Scientific
Varning
PC409518-1G PC409518-5G
APOSPC409518-1GEA 11300 SEK
APOSPC409518-1G APOSPC409518-5G
3-[4-(Trifluoromethyl)phenyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine 95+%
3-[4-(Trifluoromethyl)phenyl]-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine
Formel: C₁₃H₁₂F₃N₃
Storage Temperature: Omgivande
MDL Number: MFCD07403548
CAS nummer: 879072-54-5