4-Fluorocyclohexanone

Leverantör: Apollo Scientific
Varning

PC409038-1G PC409038-250MG
APOSPC409038-1GEA 12475 SEK
APOSPC409038-1G APOSPC409038-250MG
4-Fluorocyclohexanone
4-Fluorocyclohexanone
Formel: C₆H₉FO
MW: 116.14 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11520472
CAS nummer: 68223-64-3
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR