1,1,1-Trifluoro-2,4-hexanedione 95%
Leverantör: Apollo Scientific
|
Varning
|
PC4087-1G
PC4087-250MG
PC4087-500MG
PC4087-5G
APOSPC4087-1GEA
1400
SEK
APOSPC4087-1G
APOSPC4087-250MG
APOSPC4087-500MG
APOSPC4087-5G
1,1,1-Trifluoro-2,4-hexanedione 95%
1,1,1-Trifluoro-2,4-hexanedione
Formel:
C₆H₇F₃O₂ MW: 168.12 g/mol Kokpunkt: 125 °C Densitet: 1.22 Flampunkt: 28 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00051306 CAS nummer: 400-54-4 EINECS: 206-924-2 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...