4-Chloro-2-fluoro-6-iodoaniline 99%

Leverantör: Apollo Scientific
Varning

PC4085-5G PC4085-10G PC4085-1G
APOSPC4085-5GEA 210 SEK
APOSPC4085-5G APOSPC4085-10G APOSPC4085-1G
4-Chloro-2-fluoro-6-iodoaniline 99%
4-Chloro-2-fluoro-6-iodoaniline
Formel: C₆H₄ClFIN
MW: 271,46 g/mol
Smältpunkt: 39…41 °C
Storage Temperature: Omgivande
MDL Number: MFCD01318122
CAS nummer: 216393-67-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR