2-(2-Fluorophenyl)acetamide 98%

Leverantör: Apollo Scientific
Varning

PC408273-1G PC408273-5G
APOSPC408273-1GEA 855 SEK
APOSPC408273-1G APOSPC408273-5G
2-(2-Fluorophenyl)acetamide 98%
2-(2-Fluorophenyl)acetamide
Formel: C₈H₈FNO
Smältpunkt: 157…160 °C
Storage Temperature: Omgivande
MDL Number: MFCD11643209
CAS nummer: 326-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR