5-Iodo-1,1,1-trifluoropentane 95%

Leverantör: Apollo Scientific
Varning

PC408218-1G PC408218-5G
APOSPC408218-1GEA 415 SEK
APOSPC408218-1G APOSPC408218-5G
5-Iodo-1,1,1-trifluoropentane 95%
5-Iodo-1,1,1-trifluoropentane
Formel: C₅H₈F₃I
Kokpunkt: 153 °C
Storage Temperature: Omgivande
MDL Number: MFCD06247810
CAS nummer: 352-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR