4-tert-Butoxy-3-fluoroaniline, Technical Grade
Leverantör: Apollo Scientific
Varning
PC408076-1G PC408076-250MG
APOSPC408076-1GEA 8800 SEK
APOSPC408076-1G APOSPC408076-250MG
4-tert-Butoxy-3-fluoroaniline, Technical Grade
4-tert-Butoxy-3-fluoroaniline
Formel: C₁₀H₁₄FNO
Storage Temperature: Omgivande
MDL Number: MFCD11052316
CAS nummer: 1039941-26-8