2-Methoxy-5-(pentafluorothio)benzaldehyde 97%
Leverantör: Apollo Scientific
Varning
PC405679-1G PC405679-5G
APOSPC405679-1GEA 4100 SEK
APOSPC405679-1G APOSPC405679-5G
2-Methoxy-5-(pentafluorothio)benzaldehyde 97%
2-Methoxy-5-(pentafluorothio)benzaldehyde
Formel: C₈H₇F₅O₂S
MW: 262.19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16652487
CAS nummer: 1240256-83-0