3,3-Difluorocyclobutanamine hydrochloride 97%
Leverantör: Apollo Scientific
Varning
PC405003-1G PC405003-5G
APOSPC405003-1GEA 260 SEK
APOSPC405003-1G APOSPC405003-5G
3,3-Difluorocyclobutanamine hydrochloride 97%
3,3-Difluorocyclobutanamine hydrochloride
Formel: C₄H₈ClF₂N
MW: 143.56 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10001382
CAS nummer: 637031-93-7