(4-Fluorophenoxy)acetic hydrazide 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(4-Fluorophenoxy)acetohydrazide, (4-Fluorophenoxy)acetic acid hydrazide, (4-Fluorophenoxy)acetohydrazide
PC4050-100G PC4050-25G PC4050-5G
APOSPC4050-100GEA 3800 SEK
APOSPC4050-100G APOSPC4050-25G APOSPC4050-5G
(4-Fluorophenoxy)acetic hydrazide 98%
(4-Fluorophenoxy)acetic hydrazide
Formel: (C₆H₄F)OCH₂CONHNH₂
MW: 184.17 g/mol
Smältpunkt: 124…126 °C
Storage Temperature: Omgivande
MDL Number: MFCD00082651
CAS nummer: 1737-62-8