3,5-Bis(trifluoromethyl)cinnamic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-(3,5-Bis(trifluoromethyl)phenyl)acrylic acid
PC4038-100G PC4038-25G PC4038-5G
APOSPC4038-100GEA 1475 SEK
APOSPC4038-100G APOSPC4038-25G APOSPC4038-5G
3,5-Bis(trifluoromethyl)cinnamic acid 98%
3,5-Bis(trifluoromethyl)cinnamic acid
Formel: C₁₁H₆F₆O₂
MW: 284,1546 g/mol
Smältpunkt: 166…169 °C
Storage Temperature: Omgivande
MDL Number: MFCD00799487
CAS nummer: 155814-20-3