5-Bromo-3-fluoropyridin-2-amine 98%
Leverantör: Apollo Scientific
Synonyms: 2-Amino-5-bromo-3-fluoropyridine, 5-Bromo-3-fluoropyridin-2-amine
PC400726-1G PC400726-10G PC400726-5G
APOSPC400726-1GEA 720 SEK
APOSPC400726-1G APOSPC400726-10G APOSPC400726-5G
5-Bromo-3-fluoropyridin-2-amine 98%
5-Bromo-3-fluoropyridin-2-amine
Formel: C₅H₄BrFN₂
MW: 191,0011 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11848483
CAS nummer: 748812-37-5