(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid
Leverantör: Apollo Scientific
PC400633-1G PC400633-250MG PC400633-500MG PC400633-2.5G
APOSPC400633-1GEA 2875 SEK
APOSPC400633-1G APOSPC400633-250MG APOSPC400633-500MG APOSPC400633-2.5G
(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid
(3-Chloro-5-(trifluoromethyl)pyridin-2-yloxy)acetic acid
Formel: C₈H₅ClF₃NO₃
MW: 255.58 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10568276
CAS nummer: 95355-49-0