5-Fluoro-2-nitrophenol 99%

Leverantör: Apollo Scientific
Varning

Synonyms: 4-Fluoro-2-hydroxynitrobenzene

PC3980-250G PC3980-1KG
APOSPC3980-250GEA 1100 SEK
APOSPC3980-250G APOSPC3980-1KG
5-Fluoro-2-nitrophenol 99%
5-Fluoro-2-nitrophenol
Formel: C₆H₄FNO₃
MW: 157,1 g/mol
Kokpunkt: 260 °C
Smältpunkt: 30…37 °C
Flampunkt: 91 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007107
CAS nummer: 446-36-6
EINECS: 207-168-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR