3-Fluoro-4-nitrophenol 99%
Leverantör: Apollo Scientific
|
Fara
|
PC3976-25G
PC3976-5G
APOSPC3976-25GEA
1300
SEK
APOSPC3976-25G
APOSPC3976-5G
3-Fluoro-4-nitrophenol 99%
3-Fluoro-4-nitrophenol
Formel:
C₆H₄FNO₃ MW: 157,1 g/mol Smältpunkt: 91…95 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00041251 CAS nummer: 394-41-2 EINECS: 206-895-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...