2,6-Dichloro-5-fluoronicotinic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 2,6-Dichloro-5-fluoropyridine-3-carboxylic acid
PC3941-100G PC3941-25G PC3941-250G
APOSPC3941-100GEA 635 SEK
APOSPC3941-100G APOSPC3941-25G APOSPC3941-250G
2,6-Dichloro-5-fluoronicotinic acid 97%
2,6-Dichloro-5-fluoronicotinic acid
Formel: C₆H₂Cl₂FNO₂
MW: 209,99 g/mol
Smältpunkt: 152…155 °C
Storage Temperature: Omgivande
MDL Number: MFCD00799517
CAS nummer: 82671-06-5