2,2,2-Trifluoroethyl trifluoromethanesulphonate 97%
Leverantör: Apollo Scientific
Fara
Synonyms: 2,2,2-Trifluoroethyltrifluoromethanesulphonate
PC3903-25G PC3903-5G PC3903-100G
APOSPC3903-25GEA 325 SEK
APOSPC3903-25G APOSPC3903-5G APOSPC3903-100G
2,2,2-Trifluoroethyl trifluoromethanesulphonate 97%
2,2,2-Trifluoroethyl trifluoromethanesulphonate
Powerful trifluoroethylating agent; Useful for synthesis offluorinated amino acids, ask for references.
Formel: C₃H₂F₆O₃S
MW: 232.10 g/mol
Kokpunkt: 91 °C
Densitet: 1.611 g/cm³ (25 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00671579
CAS nummer: 6226-25-1
EINECS: 458-390-7
UN: 3265
ADR: 8 II