2-Fluoro-1-methylpyridinium-p-toluenesulphonate 95%
Leverantör: Apollo Scientific
Synonyms: 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate
PC3826-25G PC3826-5G
APOSPC3826-25GEA 2750 SEK
APOSPC3826-25G APOSPC3826-5G
2-Fluoro-1-methylpyridinium-p-toluenesulphonate 95%
2-Fluoro-1-methylpyridinium-p-toluenesulphonate
Peptide coupling agent. see: Chem.Lett.,547(1984)for synthesis of alpha-ribonucleotides
Formel: C₁₃H₁₄FNO₃S
MW: 283,32 g/mol
Smältpunkt: 132…134 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011983
CAS nummer: 58086-67-2