2,2,3,3,4,4-Hexafluoro-1,5-pentyl diacrylate 95%

Leverantör: Apollo Scientific
Varning

PC3811-5G PC3811-1G
APOSPC3811-5GEA 5750 SEK
APOSPC3811-5G APOSPC3811-1G
2,2,3,3,4,4-Hexafluoro-1,5-pentyl diacrylate 95%
2,2,3,3,4,4-Hexafluoro-1,5-pentyl diacrylate
Formel: C₁₁H₁₀F₆O₄
MW: 320,19 g/mol
Kokpunkt: 105 °C/0,1mmHg
Storage Temperature: Kylskåp
MDL Number: MFCD02093323
CAS nummer: 678-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR