Hafnium(IV) trifluoromethanesulphonate 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Trifluoromethanesulphonic acid Hafnium(IV) salt, Hafnium(IV) Triflate
PC3761-1G PC3761-5G
APOSPC3761-1GEA 1350 SEK
APOSPC3761-1G APOSPC3761-5G
Hafnium(IV) trifluoromethanesulphonate 98%
Hafnium(IV) trifluoromethanesulphonate
Formel: CHF₃O₃S
MW: 774,74 g/mol
Smältpunkt: >350 °C
Storage Temperature: Omgivande
MDL Number: MFCD01321255
CAS nummer: 161337-67-3