4-Fluoro-DL-glutamic acid 95%

Leverantör: Apollo Scientific
Varning

PC3753-100MG PC3753-1G PC3753-500MG
APOSPC3753-100MGEA 1900 SEK
APOSPC3753-100MG APOSPC3753-1G APOSPC3753-500MG
4-Fluoro-DL-glutamic acid 95%
4-Fluoro-DL-glutamic acid
Formel: C₅H₈FNO₄
MW: 165,12 g/mol
Smältpunkt: 185 °C
Storage Temperature: Omgivande
MDL Number: MFCD00055778
CAS nummer: 2708-77-2
EINECS: 220-302-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR