2-Fluorodiphenylmethane 99%

Leverantör: Apollo Scientific

PC3748E-1G PC3748E-5G
APOSPC3748E-1GEA 1600 SEK
APOSPC3748E-1G APOSPC3748E-5G
2-Fluorodiphenylmethane 99%
2-Fluorodiphenylmethane
Formel: C₁₃H₁₁F
Kokpunkt: 76 °C/1mmHg
Storage Temperature: Omgivande
MDL Number: MFCD00079758
CAS nummer: 3794-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR