Methyl-3-bromo-2-fluorobenzoate

Leverantör: Apollo Scientific
Varning

PC3663-1G PC3663-5G
APOSPC3663-1GEA 290 SEK
APOSPC3663-1G APOSPC3663-5G
Methyl-3-bromo-2-fluorobenzoate
Methyl-3-bromo-2-fluorobenzoate
Formel: C₈H₆BrFO₂
Smältpunkt: 35…37 °C
Storage Temperature: Omgivande
MDL Number: MFCD09261255
CAS nummer: 206551-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR