4-Fluorobenzyl alcohol 95+%

Leverantör: Apollo Scientific
Varning

Synonyms: (4-Fluorophenyl)methanol

PC3660-100G PC3660-25G
APOSPC3660-100GEA 1475 SEK
APOSPC3660-100G APOSPC3660-25G
4-Fluorobenzyl alcohol 95+%
4-Fluorobenzyl alcohol
Formel: C₇H₇FO
MW: 126,13 g/mol
Kokpunkt: 209 °C/730mmHg
Smältpunkt: 23 °C
Densitet: 1,156 g/cm³ (25 °C)
Flampunkt: 90 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004651
CAS nummer: 459-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR