4-Fluoro-m-anisidine hydrochloride 98%

Leverantör: Apollo Scientific

Synonyms: 4-Fluoro-3-methoxyaniline hydrochloride

PC3585-1G PC3585-5G
APOSPC3585-1GEA 1275 SEK
APOSPC3585-1G APOSPC3585-5G
4-Fluoro-m-anisidine hydrochloride 98%
4-Fluoro-m-anisidine hydrochloride
Formel: C₇H₉ClFNO
MW: 177,6000 g/mol
Smältpunkt: 248…250 °C
Storage Temperature: Kylskåp
MDL Number: MFCD08689481
CAS nummer: 22510-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR