3-Fluoro-4-(methoxy(methyl)carbamoyl)phenylboronic acid 98%
Leverantör: Apollo Scientific
Synonyms: 3-Fluoro-4-[methoxy(methyl)carbamoyl]phenylboronic acid, (3-Fluoro-4-(methoxy(methyl)carbamoyl)phenyl)boronic acid
PC3556-1G PC3556-5G
APOSPC3556-1GEA 1925 SEK
APOSPC3556-1G APOSPC3556-5G
3-Fluoro-4-(methoxy(methyl)carbamoyl)phenylboronic acid 98%
3-Fluoro-4-(methoxy(methyl)carbamoyl)phenylboronic acid
Formel: C₉H₁₁BFNO₄
MW: 227.0000 g/mol
Smältpunkt: 154…156 °C
Storage Temperature: Kylskåp
MDL Number: MFCD08689477
CAS nummer: 913835-59-3