4-Methoxy-2-(trifluoromethyl)benzoic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Methoxy-2-trifluormethylbenzoic acid
PC3498-1G PC3498-5G PC3498-25G
APOSPC3498-1GEA 395 SEK
APOSPC3498-1G APOSPC3498-5G APOSPC3498-25G
4-Methoxy-2-(trifluoromethyl)benzoic acid 98%
4-Methoxy-2-(trifluoromethyl)benzoic acid
Formel: C₉H₇F₃O₃
MW: 220,15 g/mol
Smältpunkt: 147…150 °C
Storage Temperature: Omgivande
MDL Number: MFCD01091013
CAS nummer: 127817-85-0