2,3,5-Trifluorocinnamic acid 97%

Leverantör: Apollo Scientific

PC3419-5G PC3419-1G
APOSPC3419-5GEA 1400 SEK
APOSPC3419-5G APOSPC3419-1G
2,3,5-Trifluorocinnamic acid 97%
2,3,5-Trifluorocinnamic acid
Formel: C₉H₅F₃O₂
MW: 202,13 g/mol
Storage Temperature: Omgivande
CAS nummer: 237761-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR