Perfluoro-compound FC-40™
Leverantör: Apollo Scientific
Varning
Synonyms: FluorinertR(FC-40)
PC3312-1KG PC3312-250G
APOSPC3312-1KGEA 7750 SEK
APOSPC3312-1KG APOSPC3312-250G
Perfluoro-compound FC-40™
Perfluoro-compound FC-40™
(Fluorinert is a registered trademark of 3M)
Formel: C₂₁F₄₈N₂
MW: 1192.168 g/mol
Kokpunkt: 155 °C
Smältpunkt: –57 °C
Densitet: 1.87 g/cm³ (25 °C)
Storage Temperature: Omgivande
MDL Number: MFCD01632188
CAS nummer: 51142-49-5