Ethyl-3-(trifluoromethyl)benzoate 97%

Leverantör: Apollo Scientific
Varning

Synonyms: Ethyl 3-(trifluoromethyl)benzoate

PC3287-100G PC3287-25G
APOSPC3287-100GEA 2475 SEK
APOSPC3287-100G APOSPC3287-25G
Ethyl-3-(trifluoromethyl)benzoate 97%
Ethyl-3-(trifluoromethyl)benzoate
Formel: C₁₀H₉F₃O₂
MW: 218,18 g/mol
Kokpunkt: 100…101 °C/10mmHg
Storage Temperature: Omgivande
MDL Number: MFCD00013560
CAS nummer: 76783-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR