4-Amino-3,5-dichloro-2,6-difluoropyridine, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: 3,5-Dichloro-2,6-difluoropyridin-4-amine
PC32132-5G PC32132-1G PC32132-25G
APOSPC32132-5GEA 620 SEK
APOSPC32132-5G APOSPC32132-1G APOSPC32132-25G
4-Amino-3,5-dichloro-2,6-difluoropyridine, Technical Grade
4-Amino-3,5-dichloro-2,6-difluoropyridine
Formel: C₅H₂Cl₂F₂N₂
MW: 198,99 g/mol
Smältpunkt: 112…114 °C
Storage Temperature: Omgivande
MDL Number: MFCD00011574
CAS nummer: 2840-00-8
EINECS: 220-630-1